Stampa…
Triethylaluminium 1.3 mol/l in heptane, AcroSeal®
Fornitore: Thermo Scientific
Pericolo
Synonyms: Triethylaluminum
377291000 377298000
377291000.EA 108 EUR
377291000. 377298000.
Triethylaluminium 1.3 mol/l in heptane, AcroSeal®
Triethylaluminium
Formula: C₆H₁₅Al
Peso molecolare: 114,17 g/mol
Punto di fusione: –93 °C
MDL Number: MFCD00009015
CAS Number: 97-93-8
EINECS: 202-619-3
UN: 3394
Visualizza i codici
Appearance Clear colorless Solution
Strength 19.5 to 20.5 wt % (on Al basis)