(4-Chlorobutyl)benzene ≥97%
Fornitore: Thermo Scientific
|
Attenzione
|
A11753.03
A11753.06
A11753.14
A11753.03EA
33.5
EUR
A11753.03
A11753.06
A11753.14
(4-Chlorobutyl)benzene ≥97%
(4-Chlorobutyl)benzene
Purity: 97%
CAS No.: 4830-93-7
Molecular Formula: C6H5(CH2)4Cl
Formula Weight: 168.67
Flash Point: 104°(219°F)
Refractive Index: 1.5180
Boiling Point: 125-128°/20mm
Beilstein Reference: 2042123
MDL No.: MFCD00037117
Formula:
(C₆H₅)(CH₂)₄Cl Peso molecolare: 168,67 g/mol Punto di ebollizione: 125…128 °C (20 mmHg) Densità: 1,026 Flash Pt: 104 °C (219 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00037117 CAS Number: 4830-93-7 EINECS: 225-408-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...