4-Ethylbenzoic acid ≥99%
Fornitore: Thermo Fisher Scientific
Purity: 99%
CAS No.: 619-64-7
Molecular Formula: C9H10O2
Formula Weight: 150.18
Melting Point: 111-114°
Beilstein Reference: 2041840
Formula:
C₉H₁₀O₂ Peso molecolare: 150.18 g/mol |
MDL Number:
MFCD00002570 CAS Number: 619-64-7 |