Stampa…
Accessories for analogue and digital block heaters
Fornitore: FALC INSTRUMENTS
621042312 621042310
460-0162EA 161 EUR
460-0162 460-0161
Accessories for analogue and digital block heaters
Riscaldatori a blocchi Blocchi
Interchangeable aluminium heating blocks for Thermoblock heaters.
  • Blocks have a separate well to accommodate a thermometer or probe if required
Single block W×D×H: 75×100×50 mm
Informazioni per l'ordine: Supplied with block extraction tool.