Stampa…
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Fornitore: Simport Scientific
T320-2N T320-2B T320-2G T320-2R T320-2Y T320-3B T320-3G T320-3R T320-3Y T320-3N
731-0436EA 173 EUR
731-0436 731-0434 731-0435 731-0437 731-0438 731-0442 731-0443 731-0445 731-0446 731-0444
Strips of 8 PCR tubes with attached caps, 0,2 ml, Amplitube™
Tubi Tubi per PCR
PP
  • Can be used with eight-channel hand-held pipettors
  • Choice of colourless and four non-cytotoxic and non-metallic colours
  • The cap has an integral seal preventing contamination with surface of lid
This more convenient 0,2 ml tube strip incorporates individually attached caps. No need to carry two separate components in inventory. The strip includes 8 integral 0,2 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer. This PCR reaction strip is available with either flat (needle pierceable) or dome-toppedMore Product Information