1,4,8,11-Tetraazacyclotetradecane ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
CYCLAM
A11516.03
A11516.06
A11516.14
A11516.03EA
69.4
EUR
A11516.03
A11516.06
A11516.14
1,4,8,11-Tetraazacyclotetradecane ≥98%
1,4,8,11-Tetraazacyclotetradecane
Purity: 98+%
CAS No.: 295-37-4
Molecular Formula: C10H24N4
Formula Weight: 200.33
Melting Point: 184-188°
Beilstein Reference: 111811
Formula:
C₁₀H₂₄N₄ Peso molecolare: 200,33 g/mol Punto di fusione: 184…188 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00005105 CAS Number: 295-37-4 EINECS: 206-039-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...