Stampa…
Isobutylboronic acid 98%
Fornitore: Thermo Fisher Scientific
Attenzione
Synonyms: 2-Methylpropylboronic acid, (2-Methylpropyl)boronic acid
359950010 359950050 359950250
ACRO359950010EA 27.7 EUR
ACRO359950010 ACRO359950050 ACRO359950250
Isobutylboronic acid 98%
Isobutylboronic acid
Formula: C₄H₁₁BO₂
Peso molecolare: 101.94 g/mol
Punto di fusione: 108…111 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00134156
CAS Number: 84110-40-7
Visualizza i codici
Appearance White to cream Crystalline powder or flakes
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %