1-Phenyl-1-propyne ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
1-Propynylbenzene, Prop-1-yn-1-ylbenzene
Purity: 98%
CAS No.: 673-32-5
Molecular Formula: C9H8
Formula Weight: 116.16
Storage Temperature: 2°C to 8°C
Flash Point: 62°(143°F)
Refractive Index: 1.5640
Boiling Point: 183-185°
Beilstein Reference: 1071474
MDL No.: MFCD00009272
Formula:
C₆H₅CCCH₃ Peso molecolare: 116,16 g/mol Punto di ebollizione: 183…185 °C Densità: 0,933 Flash Pt: 62 °C (143 °F) Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00009272 CAS Number: 673-32-5 EINECS: 211-607-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...