Stampa…
Pinze, a tre punte
Fornitore: BOCHEM
5420 5551
BOCH5420EA 251 EUR
BOCH5420 BOCH5551
Pinze, a tre punte
Morsetti Clamp per uso generico
Three pronged retort clamps made of stainless steel.
  • Fingers are covered with a silicone coating