1-Aminocyclopropanecarboxylic acid ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Amino-1-cyclopropanecarboxylic acid
A1178-1G
A1178-100MG
A1178-5G
TCIAA1178-1GEA
85
EUR
TCIAA1178-1G
TCIAA1178-100MG
TCIAA1178-5G
1-Aminocyclopropanecarboxylic acid ≥97.0% (by titrimetric analysis)
1-Aminocyclopropanecarboxylic acid
Formula:
C₄H₇NO₂ Peso molecolare: 101,11 g/mol |
MDL Number:
MFCD00009944 CAS Number: 22059-21-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...