Stampa…

3-Iodophenylboronic acid pinacol ester ≥97%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: 3-Iodobenzeneboronic acid pinacol ester, 2-(3-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H28115.06 H28115.03
H28115.06EA 126 EUR
H28115.06 H28115.03
3-Iodophenylboronic acid pinacol ester ≥97%
3-Iodophenylboronic acid pinacol ester

Purity: 97% CAS No.: 408492-28-4 Molecular Formula: C12H16BIO2 Formula Weight: 329.97 Melting Point: 70-72° MDL No.: MFCD09953498

Formula: C₁₂H₁₆BIO₂
Punto di fusione: 70…72 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09953498
CAS Number: 408492-28-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR