Stampa…

Isobutylboronic acid ≥97%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: 2-Methylpropylboronic acid, (2-Methylpropyl)boronic acid

H27698.06 H27698.03
H27698.06EA 88.6 EUR
H27698.06 H27698.03
Isobutylboronic acid ≥97%
Isobutylboronic acid

Purity: 97% CAS No.: 84110-40-7 Molecular Formula: (CH3)2CHCH2B(OH)2 Formula Weight: 101.94 Melting Point: 108-110° MDL No.: MFCD00134156

Formula: C₄H₁₁BO₂
Peso molecolare: 101.94 g/mol
Punto di fusione: 108…110 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00134156
CAS Number: 84110-40-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR