Methylboronic acid ≥97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 97%
CAS No.: 13061-96-6
Molecular Formula: CH5BO2
Formula Weight: 59.86
Odour: Odourless
Melting Point: 89-94°
Beilstein Reference: 1731087
MDL No.: MFCD00002105
Formula:
CH₅BO₂ Peso molecolare: 59.86 g/mol Punto di fusione: 89…94 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002105 CAS Number: 13061-96-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...