1-Hexylboronic acid ≥97%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
Hexylboronic acid
B22446.09
B22446.88
B22446.09EA
247
EUR
B22446.09
B22446.88
1-Hexylboronic acid ≥97%
1-Hexylboronic acid
Purity: 97%
CAS No.: 16343-08-1
Molecular Formula: CH3(CH2)5B(OH)2
Formula Weight: 130.00
Melting Point: 85-90°
Beilstein Reference: 1736080
Formula:
C₆H₁₅BO₂ Peso molecolare: 129,99 g/mol Punto di fusione: 85…90 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01074641 CAS Number: 16343-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...