1-Butylboronic acid 98%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Synonyms:
Butylboronic acid, n-Butylboronic acid
Formula:
C₄H₁₁BO₂ Punto di fusione: 94…96 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00002106 CAS Number: 4426-47-5 EINECS: 224-607-7 |
Risultati del test di specificazione
Appearance | White to off-white Glistening powder or flakes |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...