Stampa…

1-Butylboronic acid ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: Butylboronic acid, n-Butylboronic acid

A13725.03 A13725.06 A13725.14
A13725.03EA 27.4 EUR
A13725.03 A13725.06 A13725.14
1-Butylboronic acid ≥98%
1-Butylboronic acid

Purity: 98% CAS No.: 4426-47-5 Molecular Formula: CH3CH2CH2CH2B(OH)2 Formula Weight: 101.94 Storage Temperature: 2°C to 8°C Melting Point: 92-96° Beilstein Reference: 1733489 MDL No.: MFCD00002106

Formula: C₄H₁₁BO₂
Peso molecolare: 101,94 g/mol
Punto di fusione: 92…96 °C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00002106
CAS Number: 4426-47-5
EINECS: 224-607-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR