1-Cyanocyclopropanecarboxylic acid ≥97%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Purity: 97%
CAS No.: 6914-79-0
Molecular Formula: C5H5NO2
Formula Weight: 111.10
Melting Point: 143-149°
UN No.: 2923
MDL No.: MFCD00190651
Formula:
C₅H₅NO₂ Punto di fusione: 143…149 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00190651 CAS Number: 6914-79-0 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...