Isoamyl butyrate, Sigma-Aldrich®
Fornitore: Merck
|
Attenzione
|
M-Clarity™ quality level = MQ200
Formula:
C₉H₁₈O₂ Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00044888 CAS Number: 106-27-4 EINECS: 203-380-8 UN: 2620 ADR: 3,III |
Risultati del test di specificazione
Solubility | 0.5 g/l (50 °C) |
Melting Point | -73 °C |
Molar Mass | 158.24 g/mol |
Boiling Point | 178 °C |
Vapor Pressure | 1.1 hPa (20 °C) |
Flash Point | 59 °C |
Density | 0.88 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...