Stampa…

Didodecyl sulphoxide ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: Dodecyl sulphoxide

D2885-25G D2885-5G
TCIAD2885-25GEA 312 EUR
TCIAD2885-25G TCIAD2885-5G
Didodecyl sulphoxide ≥98.0% (by HPLC)
Didodecyl sulphoxide
Formula: C₂₄H₅₀OS
Peso molecolare: 386,73 g/mol
MDL Number: MFCD00039865
CAS Number: 2168-96-9
EINECS: 218-513-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78b93c4b28bae4d3a1c8ef85440200af01f8c3e7&vid=78b93c4b28bae4d3a1c8ef85440200af01f8c3e7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point gt. 89.0 to 93.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR