Stampa…

Ethylmalonic acid ≥97%

Fornitore: Thermo Fisher Scientific
Attenzione

B20715.06 B20715.14 B20715.22
B20715.06EA 31.1 EUR
B20715.06 B20715.14 B20715.22
Ethylmalonic acid ≥97%
Ethylmalonic acid

Purity: 98% CAS No.: 601-75-2 Molecular Formula: CH3CH2CH(CO2H)2 Formula Weight: 132.12 Melting Point: 112-114° Beilstein Reference: 774334

Formula: C₅H₈O₄
Punto di fusione: 112…114 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002668
CAS Number: 601-75-2
EINECS: 210-007-2

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR