3-Bromo-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
B3336-25G
B3336-5G
TCIAB3336-25GEA
94
EUR
TCIAB3336-25G
TCIAB3336-5G
3-Bromo-4-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
3-Bromo-4-methoxybenzoic acid
Formula:
C₈H₇BrO₃ |
MDL Number:
MFCD00020295 CAS Number: 99-58-1 EINECS: 202-768-4 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 220.0 to 225.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...