Stampa…

Propiolaldehyde diethyl acetal ≥97.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Propargylaldehyde diethyl acetal, 3,3-Diethoxypropyne, Propioaldehyde diethyl acetal, Propynal diethyl acetal, 3,3-Diethoxy-1-propyne, 3,3-Diethoxyprop-1-yne

P1173-25ML P1173-5ML
TCIAP1173-25MLEA 244 EUR
TCIAP1173-25ML TCIAP1173-5ML
Propiolaldehyde diethyl acetal ≥97.0% (by GC)
Propiolaldehyde diethyl acetal
Formula: C₇H₁₂O₂
Punto di ebollizione: 139°C
Densità: 0,8953 g/cm³ (20 °C)
Flash Pt: 32°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00009237
CAS Number: 10160-87-9
EINECS: 233-430-4
UN: 3271
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83a0577983edf06386222f1c4bf354258e9f52ea&vid=83a0577983edf06386222f1c4bf354258e9f52ea&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR