Stampa…

5-Bromo-2-chlorobenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

B3116-25G B3116-5G
TCIAB3116-25GEA 387 EUR
TCIAB3116-25G TCIAB3116-5G
5-Bromo-2-chlorobenzonitrile ≥98.0% (by GC)
5-Bromo-2-chlorobenzonitrile
Formula: C₇H₃BrClN
Peso molecolare: 216,46 g/mol
MDL Number: MFCD00672948
CAS Number: 57381-44-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a32fe10f0a3d323755de230ad6f656f90cd95464&vid=a32fe10f0a3d323755de230ad6f656f90cd95464&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR