Stampa…

2-Bromo-4-fluorobenzaldehyde ≥98.0% (by GC)

Fornitore: TCI
Attenzione

B3498-25G B3498-5G
TCIAB3498-25GEA 134 EUR
TCIAB3498-25G TCIAB3498-5G
2-Bromo-4-fluorobenzaldehyde ≥98.0% (by GC)
2-Bromo-4-fluorobenzaldehyde
Formula: C₇H₄BrFO
MDL Number: MFCD00672923
CAS Number: 59142-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36a720bfc05ad66800de04ffaa027ad06e0324d4&vid=36a720bfc05ad66800de04ffaa027ad06e0324d4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR