Oro ≥99.95% (metals basis), nanopowder APS 20-25 nm
Fornitore: Thermo Fisher Scientific
Purity: 99.95+%
CAS No.: 7440-57-5
Appearance: Gold
Odour: Odourless
MDL No.: MFCD00003436
Formula:
Au Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...