Stampa…
Water baths, analogue, multiposition, WB-S and WB-U series
Fornitore: FALC INSTRUMENTS
610102206 610101204
462-0332EA 3040 EUR
462-0332 462-0328
Water baths, analogue, multiposition, WB-S and WB-U series
Bagni Bagni ad aqua senza agitazione
Water baths made from stainless steel with 2,4 or 6 positions specifically designed for extractors, allowing different sized flasks to be heated without great heat loss. WB-S models have multiple baths insulated separately with glass fibre for simultaneous operation of each tank at different temperatures. WB-U models have one tank and a single controller with multiple positions.
  • Each tank is fitted with a filling and level device, temperature regulator with graduated scale and sensor
  • WB-U models supplied with a thermometer and drain cock
IP Protection Class according to DIN EN 60529: IP 43
Informazioni per l'ordine: Supplied with concentric ring lid sets, four rings per tank and 500 x10 mm (lר) support rods for each position.