Stampa…
Dischi per agitatore magnetico
Fornitore: KARTELL
777 778 779
KART777EA 10.1 EUR
KART777 KART778 KART779
Dischi per agitatore magnetico
Dispositivi agitatori Ancorette magnetiche
PTFE coated magnetic stirring bars with convex shape for good centring action.
  • Effective stirring action
  • Suitable for food stuff