Stampa…
Ancorette magnetiche, con anello centrale
Fornitore: KARTELL
788 787 793 797 799 794 786 0078900
KART788EA 71.8 EUR
KART788 KART787 KART793 KART797 KART799 KART794 KART786 KART0078900
Ancorette magnetiche, con anello centrale
Dispositivi agitatori Ancorette magnetiche
PTFE coated suitable for use in containers with irregular bottom.
  • Angled surfaces create high turbulence even at low speed
  • Pivot ring reduces the contact surface and fosters optimal stir bar positioning
  • Suitable for foodstuff