Stampa…

Acido 2,4,6-triclorobenzoico ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,4,6-TBA

T1518-10G T1518-25G
TCIAT1518-10GEA 62 EUR
TCIAT1518-10G TCIAT1518-25G
Acido 2,4,6-triclorobenzoico ≥98.0% (by GC, titration analysis)
Acido 2,4,6-triclorobenzoico
Formula: C₇H₃Cl₃O₂
Peso molecolare: 225,46 g/mol
MDL Number: MFCD00060699
CAS Number: 50-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab961598b2ee2d0b8a2934f44bc118a3424865ac&vid=b1973d5c44d68aae18b2504852f23ab1d61e203c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 162.0 to 167.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR