Stampa…

4-Phenoxybenzoic acid ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

A17998.14 A17998.22
A17998.14EA 79.1 EUR
A17998.14 A17998.22
4-Phenoxybenzoic acid ≥99%
4-Phenoxybenzoic acid

Purity: 99% CAS No.: 2215-77-2 Molecular Formula: C13H10O3 Formula Weight: 214.22 Melting Point: 160-164° Beilstein Reference: 2212463

Formula: C₁₃H₁₀O₃
Punto di fusione: 160…164 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002539
CAS Number: 2215-77-2
EINECS: 218-682-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR