2,4,6-Trimethoxybenzoic acid ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
L04997.06
L04997.14
L04997.06EA
94.5
EUR
L04997.06
L04997.14
2,4,6-Trimethoxybenzoic acid ≥98%
2,4,6-Trimethoxybenzoic acid
Purity: 98%
CAS No.: 570-02-5
Molecular Formula: C10H12O5
Formula Weight: 212.20
Melting Point: ca 158° dec.
Beilstein Reference: 1972294
Formula:
C₁₀H₁₂O₅ Peso molecolare: 212,2 g/mol Punto di fusione: ∼158 °C (dec.) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00016497 CAS Number: 570-02-5 EINECS: 209-325-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...