Stampa…

Ammonio benzoato ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Benzoic acid ammonium salt

30753.36 30753.A3 30753.22
30753.36EA 54.5 EUR
30753.36 30753.A3 30753.22
Ammonio benzoato ≥99%
Ammonio benzoato

Purity: 99% CAS No.: 1863-63-4 Molecular Formula: C6H5CO2NH4 Formula Weight: 139.15 Flash Point: 110°(230°F) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White to off-white Odour: Weak Melting Point: 192-198° dec. Merck Reference: 14,496 MDL No.: MFCD00013069

Formula: C₇H₉NO₂
Peso molecolare: 139.15 g/mol
Punto di fusione: 192…198 °C (dec.)
Densità: 1.26 g/cm³ (20 °C)
Flash Pt: 110 °C (230 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013069
CAS Number: 1863-63-4
EINECS: 217-468-9
Merck Index: 12,00521

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR