Stampa…

N,N-Dimetilidrazina

Fornitore: Merck
Pericolo

Synonyms: 1,1-Dimetilidrazina

M-Clarity™ quality level = MQ200

8.10408.0100 8.10408.0500
8.10408.0100EA 92.7 EUR
8.10408.0100 8.10408.0500
N,N-Dimetilidrazina
N,N-Dimetilidrazina
Formula: C₂H₈N₂
Peso molecolare: 60,1 g/mol
Temperatura di stoccaggio: Refrigerata
CAS Number: 57-14-7
EINECS: 200-316-0
UN: 1163
ADR: 6.1 (3, 8),I

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR