Dichloromethylvinylsilane ≥97%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Purity: 97%
CAS No.: 124-70-9
Molecular Formula: C3H6Cl2Si
Formula Weight: 141.07
Flash Point: 4°(39°F)
Refractive Index: 1.4300
Odour: Acrid
Melting Point: -78°
Boiling Point: 92-93°
Beilstein Reference: 1740822
UN No.: 2985
MDL No.: MFCD00000492
Formula:
C₃H₆Cl₂Si Punto di ebollizione: 92…93 °C Punto di fusione: –78 °C Densità: 1,087 Flash Pt: 4 °C (39 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00000492 CAS Number: 124-70-9 EINECS: 204-710-3 UN: 2985 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...