Oxamic acid ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
2-Amino-2-oxoacetic acid
B20344.06
B20344.14
B20344.22
B20344.06EA
31.6
EUR
B20344.06
B20344.14
B20344.22
Oxamic acid ≥98%
Oxamic acid
Purity: 98%
CAS No.: 471-47-6
Molecular Formula: H2NC(O)CO2H
Formula Weight: 89.05
Melting Point: ca 205° dec.
Beilstein Reference: 1743294
Merck Reference: 14,6917
MDL No.: MFCD00008006
Formula:
C₂H₃NO₃ Peso molecolare: 89,05 g/mol Punto di fusione: ∼ 205 °C (dec.) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008006 CAS Number: 471-47-6 EINECS: 207-443-0 Merck Index: 14,6917 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...