Stampa…
4-tert-Amylphenol 99%
Fornitore: Thermo Scientific
Pericolo
Synonyms: p-(1,1-Dimethylpropyl)phenol, 4-(2-Methyl-2-butyl)phenol, 4-tert-Pentylphenol
167330050 167331000 167332500 167335000 167330000
ACRO167330050EA 19.5 EUR
ACRO167330050 ACRO167331000 ACRO167332500 ACRO167335000 ACRO167330000
4-tert-Amylphenol 99%
4-tert-Amylphenol
Formula: C₁₁H₁₆O
Peso molecolare: 164,25 g/mol
Punto di fusione: >91 °C
Flash Pt: 111 °C
MDL Number: MFCD00002369
CAS Number: 80-46-6
EINECS: 201-280-9
UN: 2430
ADR: 8,III
Visualizza i codici
Appearance White to light yellow Coarse powder or flakes or briquettes
Infrared spectrum Conforms
Melting point ≥91 °C
GC ≥98.5 %