Stampa…
Silver trifluoroacetate 98%
Fornitore: Thermo Scientific
Attenzione
Synonyms: Silver(I) trifluoroacetate
139730050 139730250
ACRO139730050EA 58.2 EUR
ACRO139730050 ACRO139730250
Silver trifluoroacetate 98%
Silver trifluoroacetate
Formula: C₂AgF₃O₂
Peso molecolare: 220,88 g/mol
Punto di fusione: 257…260 °C
MDL Number: MFCD00013199
CAS Number: 2966-50-9
EINECS: 221-004-0
UN: 3077
Visualizza i codici
Appearance Light beige to grey Crystalline powder
Infrared spectrum Conforms
Titration with NH4SCN ≥97.5 %