Stampa…
Bottles, narrow neck, square, HDPE, with screw caps, Corning® Gosselin™
Fornitore: Corning
FC1000-52 FC1000-51 FC1000-50
GOSSFC1000-52EA 244 EUR
GOSSFC1000-52 GOSSFC1000-51 GOSSFC1000-50
Bottles, narrow neck, square, HDPE, with screw caps, Corning® Gosselin™
Bottiglie Bottiglie in plastica
Made of HDPE, translucent, with PP screw cap.
  • Graduated
  • Good chemical and temperature resistance
  • Tamper evident screw cap with shaped seal
  • Screw cap assembled