Stampa…
Accessories for Vacuum filtration apparatus, Whatman™
Fornitore: Whatman products (Cytiva)
10464103 10471101 10471700 10477600 10477601
WHAT10464103EA 158 EUR
WHAT10464103 514-0373 514-0684 514-0121 514-0122
Accessories for Vacuum filtration apparatus, Whatman™
Rampe di filtrazione
Vacuum and pressure pump: For use in the fields of microbiological quality control, analyses, medicine and production technology. The pump is used for pumping gases, taking samples (even liquids in a vacuum) and evacuating vessels.
  • AC model
  • Contamination-free pumping of air, gases and vapours
  • Extremely quiet and smooth running
  • Equipped with thermo switch and standard fuse
  • Oil-free membrane pump
  • Delivery m³/h: 3,6 L/min, vacuum: <100 mbar absolute, pressure: 4 bar, weight: 11 kg
Witt's bottle: For filtrate collection in an inserted container.More Product Information