Stampa…

1-Dodecene 93-95%

Fornitore: Thermo Scientific
Pericolo

Synonyms: Dodec-1-ene

117631000 117635000 117630025 117630100
ACRO117631000EA 24.1 EUR
ACRO117631000 ACRO117635000 ACRO117630025 ACRO117630100
1-Dodecene 93-95%
1-Dodecene
Formula: C₁₂H₂₄
Peso molecolare: 168,32 g/mol
Punto di ebollizione: 213 °C (1013 hPa)
Punto di fusione: –35 °C
Densità: 0,75 g/cm³
Flash Pt: 77 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00008961
CAS Number: 112-41-4
EINECS: 203-968-4
UN: 3082
ADR: 9,III

Visualizza i codici

Risultati del test di specificazione

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4284 to 1.4304 (20°C, 589 nm)
GC ≥99.8 %
Solubility (In water) Passes test
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Carbonyl compounds Each of acetone, formaldehyde and acetaldehyde
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0002 meq/g
KMnO4-reducing substance Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR