4-Pentenal ≥97%
Fornitore: Thermo Fisher Scientific
Purity: 97%
CAS No.: 2100-17-6
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 7°(44°F)
Refractive Index: 1.4170
Boiling Point: 101-105°
Beilstein Reference: 1734369
UN No.: 1989
MDL No.: MFCD00151841
Formula:
C₅H₈O |
MDL Number:
MFCD00151841 CAS Number: 2100-17-6 |