Stampa…

Rubrene ≥97%

Fornitore: Thermo Scientific

Synonyms: 5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene

H53507.03 H53507.06 H53507.14
H53507.03EA 93.9 EUR
H53507.03 H53507.06 H53507.14
Rubrene ≥97%
Rubrene

Purity: 97% CAS No.: 517-51-1 Molecular Formula: C42H28 Formula Weight: 532.68 Melting Point: 330-335° Beilstein Reference: 1917339

Formula: C₄₂H₂₈
Peso molecolare: 532,68 g/mol
Punto di fusione: 330…335 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003703
CAS Number: 517-51-1
EINECS: 208-242-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR