Rubrene 99%
Fornitore: Thermo Scientific
Synonyms:
5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene
132380010
132385000
ACRO132380010EA
92.4
EUR
ACRO132380010
ACRO132385000
Rubrene 99%
Rubrene
Formula:
C₄₂H₂₈ Peso molecolare: 532,68 g/mol Punto di fusione: 330…335 °C |
MDL Number:
MFCD00003703 CAS Number: 517-51-1 EINECS: 208-242-0 |
Risultati del test di specificazione
Appearance | Red Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...