Stampa…

Dodecyl isocyanate ≥98%

Fornitore: Thermo Scientific
Pericolo

Synonyms: 1-Dodecyl isocyanate

L13380.03 L13380.06
L13380.03EA 34 EUR
L13380.03 L13380.06
Dodecyl isocyanate ≥98%
Dodecyl isocyanate

Purity: 98% CAS No.: 4202-38-4 Molecular Formula: CH3(CH2)11NCO Formula Weight: 211.35 Storage Temperature: 2°C to 8°C Flash Point: 134°(273°F) Refractive Index: 1.4410 Odour: Irritating Boiling Point: 157-161°/21mm Beilstein Reference: 1774823 UN No.: 2206 MDL No.: MFCD00041744

Formula: C₁₃H₂₅NO
Peso molecolare: 211,35 g/mol
Punto di ebollizione: 157…161 °C (21 mmHg)
Densità: 0,877
Flash Pt: 134 °C (273 °F)
MDL Number: MFCD00041744
CAS Number: 4202-38-4
EINECS: 224-111-0
UN: 2206
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR