Stampa…

Sodium triacetoxyborohydride 95%

Fornitore: Thermo Fisher Scientific
Pericolo

B22060.22 B22060.14 B22060.06
B22060.22EA 176 EUR
B22060.22 B22060.14 B22060.06
Sodium triacetoxyborohydride 95%
Sodium triacetoxyborohydride

Purity: 95% CAS No.: 56553-60-7 Molecular Formula: Na(CH3CO2)3BH Formula Weight: 211.94 Melting Point: ca 110° dec. Beilstein Reference: 4047608 UN No.: 3396 MDL No.: MFCD00012211

Formula: C₆H₁₀BNaO₆
Punto di fusione: ∼ 110 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012211
CAS Number: 56553-60-7
UN: 3396
ADR: 4.3,III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR