Ethoxyacetic acid 98%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Formula:
C₄H₈O₃ Punto di ebollizione: 97…100 °C (11 mmHg) Densità: 1,102 g/cm³ Flash Pt: 97 °C |
MDL Number:
MFCD00004310 CAS Number: 627-03-2 EINECS: 210-978-2 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4170 to 1.4200 (20°C, 589 nm) |
GC | ≥97.5 % |
Residue after evaporation | ≤0.0005 % |
Acidity/alkalinity | ≤0.0005 meq/g |
Filt.over 0.2 micron PFTE | Filtered |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...