Stampa…

Ethynylcyclohexane ≥98%

Fornitore: Thermo Scientific
Pericolo

Synonyms: Cyclohexylacetylene

H53418.14 H53418.06
H53418.14EA 428 EUR
H53418.14 H53418.06
Ethynylcyclohexane ≥98%
Ethynylcyclohexane

Purity: 98% CAS No.: 931-48-6 Molecular Formula: C8H12 Formula Weight: 108.18 Flash Point: 18°(64°F) Refractive Index: 1.4540 Boiling Point: 130-132° Beilstein Reference: 1815535 UN No.: 3295 MDL No.: MFCD00001513

Formula: C₈H₁₂
Peso molecolare: 108,18 g/mol
Punto di ebollizione: 130…132 °C
Densità: 0,828
Flash Pt: 18 °C (64 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00001513
CAS Number: 931-48-6
EINECS: 213-236-6
ADR: 3,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR