Stampa…

Tributylvinyltin 96%

Fornitore: Thermo Scientific
Pericolo

Synonyms: Tri-n-butylvinyltin, Tributylvinylstannane

L20215.03 L20215.06 L20215.14
L20215.03EA 49.9 EUR
L20215.03 L20215.06 L20215.14
Tributylvinyltin 96%
Tributylvinyltin

Purity: 96% CAS No.: 7486-35-3 Molecular Formula: C14H30Sn Formula Weight: 317.11 Flash Point: 110°(230°F) Refractive Index: 1.4780 Boiling Point: 104-106°/3.5mm Beilstein Reference: 3537662 UN No.: 2788 MDL No.: MFCD00009421

Formula: C₁₄H₃₀Sn
Peso molecolare: 317,1 g/mol
Punto di ebollizione: 104…106 °C (3,5 mmHg)
Densità: 1,085
Flash Pt: >110 °C (230 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009421
CAS Number: 7486-35-3
EINECS: 231-291-4
UN: 2788
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR