3-Aminothiobenzamide 97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 97%
CAS No.: 78950-36-4
Molecular Formula: C7H8N2S
Formula Weight: 152.21
Melting Point: 127-131°
MDL No.: MFCD04973325
Formula:
C₇H₈N₂S Punto di fusione: 127…131 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD04973325 CAS Number: 78950-36-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...