2-Cyano-3-methylpyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
3-Methylpyridine-2-carbonitrile, 3-Methylpicolinonitrile, 2-Cyano-β-picoline, 2-Cyano-3-picoline
C1410-25G
C1410-5G
TCIAC1410-25GEA
53
EUR
TCIAC1410-25G
TCIAC1410-5G
2-Cyano-3-methylpyridine ≥98.0% (by GC, titration analysis)
2-Cyano-3-methylpyridine
Formula:
C₇H₆N₂ Peso molecolare: 118,14 g/mol Punto di ebollizione: 142°C Punto di fusione: 87…90 °C |
MDL Number:
MFCD00190585 CAS Number: 20970-75-6 EINECS: 244-131-3 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 84.0 to 87.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...